ContactUsLinkComponent
Beakers, graduated, tall form, PP, SP Bel-Art
Beakers, graduated, tall form, PP, SP Bel-Art
Catalog # BELAF289900000
Supplier #: F289900000
undefined
Beakers, graduated, tall form, PP, SP Bel-Art
Catalog # BELAF289900000
Supplier #: F289900000
Log in for availability
Select an Option
95 mm
Select an Option
10 ml
Select an Option
500 ml
Select an Option
137 mm
Select an Option
02
Select an Option
1
About this item
Made of PP, translucent.
  • Use with wet or dry ingredients
  • Chemical resistant and shatterproof
  • Metric gradations molded on outside wall
  • Translucent polypropylene construction
  • Heat resistant to 121ºC (250ºF); steam autoclavable
Available in a large selection of sizes, these pitchers are ideal for general laboratory use and offer well designed spouts and large open handles for easy stacking.
Frequently Bought Together