Cryogenic freezer boxes, CryoFile™, WHEATON®
Supplier: DWK Life Sciences
Jump to:
About this item
High quality chip-board, spray coated with polyethylene.
- Water repellent material allows for longer durability
- Numbering system printed on lid and bottom of the box
- Withstands cryogenic temperatures to –196 °C
- Six colours provide easy sample idenfication
Use for batching and storing cryogenic vials or ampoules. Partitions numbered from 1 to 81 for easy content identification.
12 Options Available Below
Product Details & Documents
High quality chip-board, spray coated with polyethylene.
- Water repellent material allows for longer durability
- Numbering system printed on lid and bottom of the box
- Withstands cryogenic temperatures to –196 °C
- Six colours provide easy sample idenfication
Use for batching and storing cryogenic vials or ampoules. Partitions numbered from 1 to 81 for easy content identification.
Product Family Options
CryoFile™ boxes
Product Information
- CompartmentsForColourW×D×H ext.AvailabilityPrice
CryoFile XL™ boxes
Product Information
- CompartmentsForColourW×D×H ext.AvailabilityPrice