Order Entry
Austria
ContactUsLinkComponent
 
VWR®, Bovine serum albumin (BSA)
  422361V
 :  
undefined
VWR®, Bovine serum albumin (BSA)
  422361V
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • CAS No.:
    9048-46-8
    90604-29-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Purity:
    98%
  • Colour:
    Light yellow
  • pH:
    6,8 - 7,2

 

 

A protease-free grade albumin for RIA and ELISA applications, produced by a heat-shock process in caprylic acid.

  • Protease-free
  • IgG-free
  • Moisture: 5%