Order Entry
Austria
ContactUsLinkComponent
VWR® Phosphate Buffered Saline (PBS), Low Phosphate, 10X
VWR® Phosphate Buffered Saline (PBS), Low Phosphate, 10X
Catalog # 437117K
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Phosphate Buffered Saline (PBS), Low Phosphate, 10X
Catalog # 437117K
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Description:
    Phosphate buffered saline (PBS), low phosphate, 10X

Specifications

About this item

Irradiated concentrate of PBS, suitable for use in washing cells after dilution.

  • KCl 2,0 g/L
  • NaCl 70,1 g/L
  • Na₂HPO₄, 2H₂0 12,8 g/L
  • NaH₂PO₄, 1H₂O 4,4 g/L

Add 1 ml of 10X PBS to 9 ml of water to give a 1X PBS solution.

pH (20 °C): 6,90 to 7,30