qScript™ One-Step qRT-PCR kits
About this item
Synthesis of cDNA and PCR amplification are carried out in the same tube without opening between procedures. These kits are optimised to deliver maximum RT-PCR efficiency and specificity, enabling unbiased co-amplification of low copy transcripts in the presence of higher copy reference genes. As a result the qScript™ One-Step qRT-PCR Kits offer improved sensitivity over other commercially available qPCR reagents. The qScript™ One-Step Master Mix maximises the activities of both reverse transcriptase and PCR enzymes, while minimising primer-dimer formation. High specificity is assured with AccuStart™ Taq DNA Polymerase, which is bound by monoclonal antibodies for superior hot-start. Upon heating to 95 °C, the antibodies denature irreversibly releasing fully active and unmodified Taq DNA polymerase. qScript™ One-Step qRT-PCR Kits are available for all Real-Time PCR instrument platforms, including those requiring normalisation with ROX reference dye.
- Broad dynamic range ensures reliable data
- Highly sensitive for low copy RNA quantification
- Unbiased duplex amplification for more accurate results
- Simple, fast protocol with less variability
3 Options Available Below
Product Details & Documents
Synthesis of cDNA and PCR amplification are carried out in the same tube without opening between procedures. These kits are optimised to deliver maximum RT-PCR efficiency and specificity, enabling unbiased co-amplification of low copy transcripts in the presence of higher copy reference genes. As a result the qScript™ One-Step qRT-PCR Kits offer improved sensitivity over other commercially available qPCR reagents. The qScript™ One-Step Master Mix maximises the activities of both reverse transcriptase and PCR enzymes, while minimising primer-dimer formation. High specificity is assured with AccuStart™ Taq DNA Polymerase, which is bound by monoclonal antibodies for superior hot-start. Upon heating to 95 °C, the antibodies denature irreversibly releasing fully active and unmodified Taq DNA polymerase. qScript™ One-Step qRT-PCR Kits are available for all Real-Time PCR instrument platforms, including those requiring normalisation with ROX reference dye.
- Broad dynamic range ensures reliable data
- Highly sensitive for low copy RNA quantification
- Unbiased duplex amplification for more accurate results
- Simple, fast protocol with less variability
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 733-1257
- 733-1259
- 733-1261
- Application:
- RT-qPCR probe
- RT-qPCR probe
- RT-qPCR probe
- One step:
- Yes
- Yes
- Yes
Specifications
Product Family Options
qScript™ One-Step qRT-PCR Kits, without passive reference dye
Product Information
- Cycler compatibilityPkAvailabilityPrice
- 733-1257qScript™ One-Step qRT PCR Kit, without passive reference dye, 200 reactionsqScript™ One-Step qRT PCR Kit, without passive reference dye, 200 reactionsAgilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480200 AssaysLog in for availability687,00 €Specifications:
More
Specifications
Cat. No.733-1257ApplicationRT-qPCR probeOne stepYes
qScript™ One-Step qRT-PCR Kits, ROX
Product Information
- Cycler compatibilityPkAvailabilityPrice
- 733-1259qScript™ One-Step qRT PCR Kit, with ROXqScript™ One-Step qRT PCR Kit, with ROXAgilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480200 AssaysLog in for availability687,00 €Specifications:
More
Specifications
Cat. No.733-1259ApplicationRT-qPCR probeOne stepYes
qScript™ One-Step qRT-PCR Kits, low ROX
Product Information
- Cycler compatibilityPkAvailabilityPrice
- 733-1261qScript™ One-Step qRT PCR Kit, with low ROXqScript™ One-Step qRT PCR Kit, with low ROXAgilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480200 AssaysLog in for availability815,00 €Specifications:
More
Specifications
Cat. No.733-1261ApplicationRT-qPCR probeOne stepYes