Orders LinkContactUsLinkComponent
Multi-way stopcocks, Discofix®
Multi-way stopcocks, Discofix®
 :   B. Braun
Multi-way stopcocks, Discofix®
 :  B. Braun
Multi-way stopcocks, Discofix®
Multi-way stopcocks, Discofix®
Suitable for all pressure infusion systems <2 bar.
  • Continuous flow channels for accurate pressure monitoring
  • Swivel lock can be moved axially or radially for fast and secure connection
  • PVC and latex-free
Suitable for all pressure infusion systems <2 bar.
Product Family Options
Discofix®-2, two-way stopcock. Handle can be rotated through 180°
  • Description
    Colour
  • Two-way stopcocks
    Blue
    1 * 50 ST
    1 * 50 ST - 145,00 €
    145,00 €
    1 * 50 ST
       145,00 €
Discofix®-3, three-way stopcock. Handle can be rotated through 360°
  • Description
    Colour
  • Three-way stopcocks
    Red
    1 * 50 ST
    1 * 50 ST - 96,70 €
    96,70 €
    1 * 50 ST
       96,70 €
  • Three-way stopcocks with 250 mm extension
    1 * 50 ST
    1 * 50 ST - 246,00 €
    246,00 €
    1 * 50 ST
       246,00 €
  • Three-way stopcocks with 1000 mm extension
    1 * 50 ST
    1 * 50 ST - 760,00 €
    760,00 €
    1 * 50 ST
       760,00 €
  • Three-way stopcocks
    Blue
    1 * 50 ST
    1 * 50 ST - 118,00 €
    118,00 €
    1 * 50 ST
       118,00 €
  • Three-way stopcocks with 100 mm extension
    1 * 50 ST
    1 * 50 ST - 246,00 €
    246,00 €
    1 * 50 ST
       246,00 €
  • Three-way stopcocks with 500 mm extension
    1 * 50 ST
    1 * 50 ST - 246,00 €
    246,00 €
    1 * 50 ST
       246,00 €
  • Three-way stopcocks
    White
    1 * 50 ST
    1 * 50 ST - 153,00 €
    153,00 €
    1 * 50 ST
       153,00 €