ContactUsLinkComponent
Infusion filter, Sterifix® Paed
Infusion filter, Sterifix® Paed
Catalog # 720-2580
Supplier:  B. Braun
undefined
Infusion filter, Sterifix® Paed
Catalog # 720-2580
Supplier:  B. Braun
Specifications
  • Colour:
    Transparent
  • Description:
    Infusion filters
  • Pk:
    50
Specifications
About this item
Sterifix Paed is specially designed for the bacteria and particle retention.
  • Reliable air venting regardless of position
  • Flow rate (aqua dest.) over 10 ml per minute
  • Luer Lock fitting