ContactUsLinkComponent
Transfer spike for sterile liquids, Transofix®
Transfer spike for sterile liquids, Transofix®
Catalog # 720-2571
Supplier:  B. Braun
undefined
Transfer spike for sterile liquids, Transofix®
Catalog # 720-2571
Supplier:  B. Braun
Specifications
  • Description:
    Transfer Spike for Sterile Liquids for bottles up to 100 ml
  • Pk:
    50
Specifications
About this item
Transofix® Transfer Set is ideal for transferring sterile liquids.
  • Conveniently designed large grip plates
  • Free of latex and PVC
  • Large internal diameter