ContactUsLinkComponent
Precision I.V. flow regulator for gravity infusions, Exadrop®
Precision I.V. flow regulator for gravity infusions, Exadrop®
Catalog # 720-2503
Supplier:  B. Braun
undefined
Precision I.V. flow regulator for gravity infusions, Exadrop®
Catalog # 720-2503
Supplier:  B. Braun
Specifications
  • Description:
    Precision I.V. flow regulator for gravity infusions, 180 cm
  • Pk:
    1
Specifications
About this item
Ready-to-use IV administration set serially equipped with precision flow regulator. Precision flow control device with or without IV administration set. Only for gravity infusion.
  • Regulator with precise graduation (ml/h)
  • Constant drip rate, independent of tubing
  • Slide clamp for short-term interruption of infusion
  • Lock fitting