Order Entry
Austria
ContactUsLinkComponent
Fine dosage syringes, Injekt®-F Solo
Fine dosage syringes, Injekt®-F Solo
Artikelnr. 720-2561
Lieferant:  B. Braun
undefined
Fine dosage syringes, Injekt®-F Solo
Artikelnr. 720-2561
Lieferant:  B. Braun

Spezifikationen

  • Capacity:
    1 ml
  • Description:
    Without needle
  • Graduation:
    0,01 ml
  • Pk:
    100

Spezifikationen

Über diesen Artikel

PP/PE fine dosage syringes, 1 ml, green plunger, two-piece, e.g. for tuberculin, heparin and allergy testing. Smear-resistant, black graduation for ideal readability. Latex and PVC free.

  • Highly transparent barrel
  • With displacement spike - no dead space, no medication wasted
  • Minimal residual volume
  • Secure plunger stopper for trouble-free filling to maximum volume
  • Centric Luer fitting
Packaging: Dispenser box of 100 units.