Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Tissue processing/embedding cassettes, Unisette™, M505

Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505
Supplier:  Simport Scientific
Certificates
Tissue processing/embedding cassettes, Unisette™, M505
Tissue processing/embedding cassettes, Unisette™, M505

11 Options Available Below

Product Details & Documents

Made from high density polymer. These cassettes, with lids, keep specimens safe whilst submerged in liquid.

  • Flow-through slots maximise fluid exchange and ensure effective drainage
  • Resistant to histological solvents
  • Integral lid
  • Front writing face inclined 35°
  • Available in 11 non cytotoxic, non metallic colours, as well as three fluorescent colours

 : Each case contains three boxes of 500 cassettes.

Product Family Options

Product Information

  • Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Pale orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Cream
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Grey
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Purple
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €

  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 231,00 €
    231,00 €
    1 * 1.500 ST
       231,00 €