Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Swingsette®, M517

Tissue processing/embedding cassettes, Swingsette®, M517

Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M517
Supplier:  Simport Scientific
Certificates
Tissue processing/embedding cassettes, Swingsette®, M517
Tissue processing/embedding cassettes, Swingsette®, M517

11 Options Available Below

Product Details & Documents

Acetal polymer.

  • Hinged lid supplied in separate packaging to make it easier to use the cassettes with commercial printers; large tab to make opening easier
  • Disposable plastic cassettes are highly resistant to histological solvents
  • Oblong opening for good fluid exchange
  • Front face inclined 45° so they can be used with certain automatic labelling systems

Packaging: Each case contains two dispenser boxes of 500 cassettes each and one dispenser box of 1000 lids.

Product Family Options

Product Information

  • Colour
    Availability
    Price
  • Purple
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • Orange
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • Salmon
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • Cream
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

  • Grey
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 159,00 €
    159,00 €
    1 * 1.000 ST
       159,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.