Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Swingsette®, M515

Tissue processing/embedding cassettes, Swingsette®, M515

Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M515
Supplier:  Simport Scientific
Certificates
Tissue processing/embedding cassettes, Swingsette®, M515
Tissue processing/embedding cassettes, Swingsette®, M515

11 Options Available Below

Product Details & Documents

Acetal polymer.

  • Integral, hinged lid, with large tab for easy opening
  • Disposable plastic cassettes highly resistant to histological solvents
  • Flow-through slots maximise fluid exchange
  • Front face inclined 45° to enable use with leading automatic labelling systems.

Packaging: Each case contains three boxes of 500 cassettes.

Product Family Options

Product Information

  • Colour
    Availability
    Price
  • Purple
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • Orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • White
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • Salmon
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • Cream
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

  • Grey
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 258,00 €
    258,00 €
    1 * 1.500 ST
       258,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.