Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Slimsette®, M510
Biopsy processing/embedding cassettes, Slimsette®, M510
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Slimsette®, M510
Supplier:  Simport Scientific
Certificates
Biopsy processing/embedding cassettes, Slimsette®, M510
Biopsy processing/embedding cassettes, Slimsette®, M510
11 Options Available Below
Product Details & Documents
Acetal polymer. Cassette design is similar to that of the M509 model but specially designed to hold biopsy specimens.
  • Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
  • Disposable plastic cassettes are highly resistant to histological solutions
  • Small, square openings for good fluid exchange
  • Integral, recessed lid
  • Front face inclined 45° to enable use with certain automatic labelling systems
W×D×H: 41×28,5×6 mm
Packaging: Each case contains three boxes holding 500 cassettes each.
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • Grey
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • Cream
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • Purple
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • Turquoise
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • Pink
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • Green
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • Yellow
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • Blue
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €
  • Salmon
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €