Orders LinkContactUsLinkComponent
Staining systems for microscope slides, DrainRack Junior
Staining systems for microscope slides, DrainRack Junior
Supplier:   Simport Scientific
Staining systems for microscope slides, DrainRack Junior
Supplier:  Simport Scientific
Certificates
Staining systems for microscope slides, DrainRack Junior
Staining systems for microscope slides, DrainRack Junior
5 Options Available Below
Product Details & Documents
This rugged tray used as a drain rack can hold up to 100 microscope slides in 50 individually numbered slots. All slides are stored upright for easier insertion and removal.
  • Barcodes can be read without having to remove the slides from the box
  • To save space, the number of slides in each slot can be doubled simply by inserting two slides in each slot
  • Not autoclavable
Product Family Options
Product Information
  • Colour
    No. of slides
    Availability
    Price
  • 631-1918
    Staining systems
    Staining systems
    White
    100
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 72,10 €
    72,10 €
    1 * 10 ST
       72,10 €
  • 631-1917
    Staining systems
    Staining systems
    Pink
    100
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 72,10 €
    72,10 €
    1 * 10 ST
       72,10 €
  • 631-1919
    Staining systems
    Staining systems
    Yellow
    100
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 72,10 €
    72,10 €
    1 * 10 ST
       72,10 €
  • 631-1915
    Staining systems
    Staining systems
    Blue
    100
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 72,10 €
    72,10 €
    1 * 10 ST
       72,10 €
  • 631-1916
    Staining systems
    Staining systems
    Green
    100
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 72,10 €
    72,10 €
    1 * 10 ST
       72,10 €