Orders LinkContactUsLinkComponent
Bath fluids
Supplier:   JULABO GmbH
Bath fluids
Supplier:  JULABO GmbH
Certificates
Bath fluids
24 Options Available Below
Product Details & Documents
Bath fluids have been carefully selected and proven in long-term service. They are suitable for use with heating and refrigerated circulators whenever water cannot be employed as bath fluid. The choice of bath fluid is critical for best results in temperature control and operation of the circulator.
  • Low toxicity and almost odourless
  • High stability and good thermal conductivity
  • Low viscosity and low corrosiveness
Product Family Options
For refrigerated circulators
Product Information
  • Type
    Temperature range
    Fire point
    Flash point
    Colour
    Pk
    Availability
    Price
  • Thermal HY
    −80...+55 °C
    80 °C
    78 °C
    Clear
    5 l
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 410,00 €
    410,00 €
    1 * 5 L
       410,00 €
  • Thermal HY
    −80...+55 °C
    80 °C
    78 °C
    Clear
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 820,00 €
    820,00 €
    1 * 10 L
       820,00 €
  • Thermal H5
    −50...+105 °C
    142 °C
    124 °C
    Clear
    5 l
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 435,00 €
    435,00 €
    1 * 5 L
       435,00 €
  • Thermal H5
    −50...+105 °C
    142 °C
    124 °C
    Clear
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 870,00 €
    870,00 €
    1 * 10 L
       870,00 €
  • Thermal G
    −30...+80 °C
    - °C
    - °C
    Light yellow
    5 l
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 123,00 €
    123,00 €
    1 * 5 L
       123,00 €
  • Thermal G
    −30...+80 °C
    - °C
    - °C
    Light yellow
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 246,00 €
    246,00 €
    1 * 10 L
       246,00 €
  • Thermal H10
    −20...+180 °C
    216 °C
    190 °C
    Clear
    5 l
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 435,00 €
    435,00 €
    1 * 5 L
       435,00 €
  • Thermal H10
    −20...+180 °C
    216 °C
    190 °C
    Clear
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 870,00 €
    870,00 €
    1 * 10 L
       870,00 €
  • Thermal H20S
    0...220 °C
    274 °C
    230 °C
    Light brown
    5 l
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 450,00 €
    450,00 €
    1 * 5 L
       450,00 €
  • Thermal H20S
    0...220 °C
    274 °C
    230 °C
    Light brown
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 900,00 €
    900,00 €
    1 * 10 L
       900,00 €
  • Thermal HS
    50...250 °C
    360 °C
    270 °C
    Light brown
    5 l
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 345,00 €
    345,00 €
    1 * 5 L
       345,00 €
  • Thermal HS
    50...250 °C
    360 °C
    270 °C
    Light brown
    10 l
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 690,00 €
    690,00 €
    1 * 10 L
       690,00 €
  • Thermal M
    70...170 °C
    306 °C
    284 °C
    Translucent
    5 l
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 265,00 €
    265,00 €
    1 * 5 L
       265,00 €
  • Thermal M
    70...170 °C
    306 °C
    284 °C
    Transparent
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 530,00 €
    530,00 €
    1 * 10 L
       530,00 €
For highly dynamic temperature control systems (Presto, Magnum and W92)
Product Information
  • Type
    Temperature range
    Fire point
    Flash point
    Colour
    Pk
    Availability
    Price
  • Thermal HL60
    −60...+250 °C
    142 °C
    124 °C
    Clear
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 450,00 €
    450,00 €
    1 * 5 L
       450,00 €
  • Thermal HL60
    −60...+250 °C
    142 °C
    124 °C
    Clear
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 900,00 €
    900,00 €
    1 * 10 L
       900,00 €
  • Thermal HL90
    −90...+200 °C
    126 °C
    >+82 °C
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 820,00 €
    820,00 €
    1 * 5 L
       820,00 €
  • Thermal HL90
    −90...+200 °C
    126 °C
    >+82 °C
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 1 640,00 €
    1 640,00 €
    1 * 10 L
       1 640,00 €
  • Thermal HL30
    −30...+90 °C
    - °C
    - °C
    Light yellow
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 128,00 €
    128,00 €
    1 * 5 L
       128,00 €
  • Thermal HL30
    −30...+90 °C
    - °C
    - °C
    Light yellow
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 256,00 €
    256,00 €
    1 * 10 L
       256,00 €
For high temperature circulators (Forte HT closed systems)
Product Information
  • Type
    Temperature range
    Fire point
    Flash point
    Colour
    Pk
    Availability
    Price
  • Thermal H250S
    20...250 °C
    274 °C
    230 °C
    Light brown
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 450,00 €
    450,00 €
    1 * 5 L
       450,00 €
  • Thermal H250S
    20...250 °C
    274 °C
    230 °C
    Light brown
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 900,00 €
    900,00 €
    1 * 10 L
       900,00 €
  • 40...300 °C
    255 °C
    245 °C
    Clear
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 390,00 €
    390,00 €
    1 * 5 L
       390,00 €
  • Thermal H350
    50...350 °C
    235 °C
    210 °C
    Clear
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 440,00 €
    440,00 €
    1 * 5 L
       440,00 €