Orders LinkContactUsLinkComponent
VWR® Surface Sampling Sponge Kits
VWR® Surface Sampling Sponge Kits
Supplier:  VWR Chemicals
Certificates
VWR® Surface Sampling Sponge Kits
VWR® Surface Sampling Sponge Kits
6 Options Available Below
Product Details & Documents
Surface sampling sponges in two forms: Dry sponge sterile sampling bag with write-on strip, containing a dry, biocide-free cellulose sponge, or wet sponge sterile sampling bag, with a foil-sealed, pre-moistened cellulose sponge, and 10 ml of sterile neutralising buffer.
  • Sterilised using gamma irradiation and leakproof transport bags, for the rapid cooling of samples in refrigerators
  • Puncture-proof safety bags eliminating sharp points that can cause puncture and/or abrasions
  • Larger surface in comparison with normal sampling swabs
This surface sampling sponge kit is designed to collect and transport samples, to detect the presence of microbiological contaminations such as Listeria, Salmonella, E. coli, and other food-borne pathogens on almost any surface. These kits are widely used in the food, medical, public health and cosmetic industries.
 : HACCP, USDA, ACIA and CFIA requirements.
 : Gloves should be worn during all sampling procedures.
Product Family Options
Product Information
  • Availability
    Price
  • 300-0229
    Sampling sponges with buffer
    Sampling sponges with buffer
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 410,00 €
    410,00 €
    1 * 100 ST
       410,00 €
  • 710-1020
    Dehydrated surface sampling sponges in a 450 ml bag
    Dehydrated surface sampling sponges in a 450 ml bag
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 792,00 €
    792,00 €
    1 * 400 ST
       792,00 €
  • 300-0230
    Surface sampling sponges pre-moistened with neutralising buffer
    Surface sampling sponges pre-moistened with neutralising buffer
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 410,00 €
    410,00 €
    1 * 100 ST
       410,00 €
  • 300-0231
    Surface sampling sponges pre-moistened with Dey/Engley neutralising buffer
    Surface sampling sponges pre-moistened with Dey/Engley neutralising buffer
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 389,00 €
    389,00 €
    1 * 100 ST
       389,00 €
  • 300-0232
    Sterile transport bags and surface sampling sponges with handle pre-moistened with neutralising buffer
    Sterile transport bags and surface sampling sponges with handle pre-moistened with neutralising buffer
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 473,00 €
    473,00 €
    1 * 100 ST
       473,00 €
  • 300-0240
    Sterile transport bags and surface sampling sponges with handle pre-moistened with Dey/Engley neutralising buffer
    Sterile transport bags and surface sampling sponges with handle pre-moistened with Dey/Engley neutralising buffer
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 452,00 €
    452,00 €
    1 * 100 ST
       452,00 €