ContactUsLinkComponent
Liquid detergent, L900, Nalgene®
Liquid detergent, L900, Nalgene®
Catalog # 148-0114
   Low Manufacturing Impact
undefined
Liquid detergent, L900, Nalgene®
Catalog # 148-0114
   Low Manufacturing Impact
Specifications
  • Description:
    Liquid detergent, L900
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    4 L
Specifications
About this item
Specifically formulated to clean plastic and glass labware used in pharmaceutical, medical research, academic, biotechnology and other facilities.
  • Formulated to be compatible with all plastics, especially polycarbonate
  • Will not cause crazing, stress cracking or clouding when used as directed
  • For hand or machine use, it is non toxic and low foaming
Non inhibitory according to APHA Standard Methods for the Examination of Water and Wastewater.