Orders LinkContactUsLinkComponent
CellStor Pot™ specimen container
Supplier:   CELLPATH
CellStor Pot™ specimen container
Supplier:  CELLPATH
Certificates
CellStor Pot™ specimen container
CellStor Pot™ specimen container
4 Options Available Below
Product Details & Documents
Prefilled specimen containers.
  • Leak-proof lid for safe transportation and specimen storage
  • With traceable badge number, safety data sheet in the packaging
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • CLLPBAF-6000-08A
    10% neutral buffered formalin
    10% neutral buffered formalin
    60 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 16,90 €
    16,90 €
    1 * 25 ST
       16,90 €
  • 720-2323
    10% formal saline
    10% formal saline
    60 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST
  • 720-2324
    10% neutral buffered formalin
    10% neutral buffered formalin
    90 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 24,60 €
    24,60 €
    1 * 25 ST
       24,60 €
  • 720-2325
    10% formal saline
    10% formal saline
    90 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 25,60 €
    25,60 €
    1 * 25 ST
       25,60 €