ContactUsLinkComponent
OCT cryo embedding matrix
OCT cryo embedding matrix
Catalog # 720-2356
Supplier:  CELLPATH
CAS Number:  
undefined
OCT cryo embedding matrix
Catalog # 720-2356
Supplier:  CELLPATH
CAS Number:  
Specifications
  • Pack type:
    Dispenser bottle
  • Pk:
    125 mL
  • Description:
    Cryotomy embedding matrix
Specifications
About this item
‘OCT’ cryo microtomy embedding medium is designed for use at –20 ˚C to –30 ˚C. The medium freezes rapidly with minimal damage by crystal formation and is an ideal support medium for cutting tissue sections.
  • Liquid level clearly visible in bottle
  • Excellent quality, highly viscous
  • Easy to use dispenser bottle
‘OCT’ is easily removed from the microscope slide prior to staining. The 125 ml bottle is a convenient size with a nozzle moulded into the cap.