Joint grease, K.W.S.
291-1310
: POLLATH
- Description:Joint vacuum and high vaccum grease, K.W.S., silicone-free
- Pk:30 mL
Conceived as vacuum and high vacuum grease. Silicon-free, no silicon derivates. No polymerisation, no sticking. Excellent adhesion and viscosity. Locked joint pairs loosened by short warming. Glass repairs are possible. Not hazardous to health in normal use.
- Scope of application: 40 to 200 °C
- Resistant to acids and lyes
- Vapour pressure: 0,00001 mbar