Forceps, ESD-safe
Conductive Vestamid with 25% fibre glass reinforcement, black. Precise, robust and virtually unbreakable forceps. Very precise; when pressure is applied during work, the tips don’t open.
- Electrically conductive
- Very high chemical resistance to acids and bases
- Heat resistant up to 175 °C
DIN 53482
Certifications: DIN 53482
Conductive Vestamid with 25% fibre glass reinforcement, black. Precise, robust and virtually unbreakable forceps. Very precise; when pressure is applied during work, the tips don’t open.
- Electrically conductive
- Very high chemical resistance to acids and bases
- Heat resistant up to 175 °C
DIN 53482
Certifications: DIN 53482
Product Family Options
- DescriptionTypeColourLength
- Straight, extra fine reinforced tipsNK7Black120 mm1 * 1 ST17,70 €