Orders LinkContactUsLinkComponent
Stirring spatulas
Stirring spatulas
Supplier:   BOETTGER PAUL
Stirring spatulas
Supplier:  BOETTGER PAUL
Certificates
Stirring spatulas
Stirring spatulas
2 Options Available Below
Product Details & Documents
PS, transparent. Stirring rod for a wide variety of laboratory applications.
  • With or without mixing plate at one end
These spatulas can be used for various applications, such as mixing samples and solutions in test tubes, adding small reagent volumes and releasing blood clots from the side of test tube walls.
Product Family Options
Product Information
  • Length
    Blade width
    Colour
    Availability
    Price
  • 231-2431
    Standard
    Standard
    120 mm
    3 mm
    Transparent
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 18,70 €
    18,70 €
    1 * 500 ST
       18,70 €
  • BOET09.251.0100
    With mixing plate
    With mixing plate
    120 mm
    3 mm
    Transparent
    Log in for availability
    Log in for availability
    1 * 6.000 ST
    1 * 6.000 ST - 259,00 €
    259,00 €
    1 * 6.000 ST
       259,00 €