Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tongue Depressors
Tongue depressor
Tongue depressor
231-6974
:
BOETTGER PAUL
Tongue depressor
231-6974
:
BOETTGER PAUL
Colour:
White
Length:
150 mm
Pk:
100
White PS.