Orders LinkContactUsLinkComponent
Specimen containers, disposable
Specimen containers, disposable
Supplier:   Simport Scientific
Specimen containers, disposable
Supplier:  Simport Scientific
Certificates
Specimen containers, disposable
Specimen containers, disposable
8 Options Available Below
Product Details & Documents
These containers and closures are made of PP. They are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples.
  • Disposable
  • Resist temperatures from −20 to + 80 °C
Tight-fitting lids prevent leaks and odors. Certain models listed below have a tight-fitting lid. Also offered with an incorporate tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover.
The 2600 and 5700 ml sizes are provided with a plastic handle to facilitate handling.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Product Family Options
Container with snap cap
Product Information
  • Capacity
    Height
    Ø opening
    Availability
    Price
  • 300 ml
    45 mm
    111 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 179,00 €
    179,00 €
    1 * 100 ST
       179,00 €
  • 500 ml
    75 mm
    111 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 164,00 €
    164,00 €
    1 * 100 ST
       164,00 €
  • 1 L
    140 mm
    111 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 239,00 €
    239,00 €
    1 * 100 ST
       239,00 €
Container, tamperproof lid
Product Information
  • Capacity
    Height
    Ø opening
    Availability
    Price
  • 300 ml
    45 mm
    111 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 193,00 €
    193,00 €
    1 * 100 ST
       193,00 €
  • 500 ml
    76 mm
    111 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 169,00 €
    169,00 €
    1 * 100 ST
       169,00 €
  • 1000 ml
    140 mm
    111 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 257,00 €
    257,00 €
    1 * 100 ST
       257,00 €
  • 2600 ml
    124 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 160,00 €
    160,00 €
    1 * 25 ST
       160,00 €
  • 5700 ml
    195 mm
    225 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 130,00 €
    130,00 €
    1 * 10 ST
       130,00 €