ContactUsLinkComponent
Measuring beaker with handle and lid
  VITL978081
 :  VITLAB
 : 978081
undefined
Measuring beaker with handle and lid
  VITL978081
 :  VITLAB
 : 978081
Select an Option
150 mm
Select an Option
50 ml
Select an Option
2000 ml
Select an Option
PP, molded, blue embossed scale
Select an Option
220 mm
Select an Option
03
Select an Option
6
Collection vessel, made of SAN, clear or PP, translucent, for a wide variety of uses.
  • Embossed scale
  • With stable, easy-grip handle
  • White PC lid
Frequently Bought Together