Disinfectant spray, Cutasept-F
: Bode
Ready to use, colourless spray propanol-based solution for rapid and gentle disinfection of skin. Bactericidal, fungicidal, tuberculocidal and viruscidal (including against enveloped viruses (incl. HCV, HIV, HBV)and non enveloped viruses eg Rotavirus*.
Composition: 100 g of solution contain the following active substances: Propan-2-ol 63,0 g,( 72 vol.%). Also contains: Benzalkonium chloride, eau purifiée.
Areas of application: Skin disinfection before injection, skin punctures ( blood sampling) and surgical procedures. Hygienic and surgical disinfection of hands.
Flash point (acc. to DIN 51755) 21 °C
: Do not use near eyes. Do not use in case of hypersensitivity to any component. Proper use of Cutasept F during pregnancy and lactation is allowed.
Side effects: Mild, diffuse skin irritation are common. Contact Allergic reactions can occur.
Side effects: Mild, diffuse skin irritation are common. Contact Allergic reactions can occur.
Frequently Bought Together