Disinfectant Solutions, Helipur H Plus N
: B. Braun
Concentrated disinfectant for temperature-sensitive instruments, endoscopes, in metal, glass or ceramic. Bactericide (inc. TbB), fungicide and virucide (hepatitis B, HIV retrovirus, etc.). Application concentration: 1,5%/60 min; 2,5%/15 min.
It is advisable to read the package leaflet before using this product.
: Helipur H Plus is corrosive.
Frequently Bought Together