Hand disinfectants, Sterillium®
Catalog # BODL9804100
Supplier: Bode
Specifications
- Description:Hand disinfectant, Sterillium®
- Pk:100 mL
Specifications
About this item
The propanol based hand disinfectant is ideal for use in all areas of the health industry (operating theatres) and hygiene sensitive industries.
- Comprehensively active against bacteria, fungi and enveloped viruses
- Protects against infections in public facilities and when traveling
- Moisturising, kind to skin with excellent skin tolerance even with long term use
- Easy to use, rub-in product with surgical standard disinfection in 90 seconds
Caution: Use biocides safely. Always read the label and product information before use.
* These articles are only available in Germany.
* These articles are only available in Germany.