Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Flat canisters

Supplier:   S.C.A.T.
Flat canisters
Supplier:  S.C.A.T.
Certificates
Flat canisters

2 Options Available Below

Product Details & Documents

Due to their flat design, SCAT flat canisters have been specially developed for use in confined spaces such as cupboards, shelves, drawers or refrigerators, as they are structurally low in height.

  • Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in laboratories
  • Available with B63 or S70/71 threads and can be adapted to many other thread sizes by means of an adapter
  • Available either as compact 8 L canister or as more voluminous 20 L canister

SCAT flat canisters are an ideal way of not having to dispense with safe storage even when space is limited, as they can be integrated into laboratory furniture to save space.

SCAT flat pack canisters are also optimally available with UN-Y approval for the storage of hazardous goods or with an integrated drain cock for safe emptying.

Product Family Options

Product Information

  • Capacity
    Colour
    Thread
    Type
    W×D×H
    Availability
    Price
  • SCTV107713
    No stopcock
    No stopcock
    20 L
    Natural
    S70/71
    UN-Y approval
    283×376×292 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,35 €
    30,35 €
    1 * 1 ST
       30,35 €

  • 8 L
    Natural
    B63
    With drain valve, autoclavable
    375×305×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 198,00 €
    198,00 €
    1 * 1 ST
       198,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.