Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M2
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M2
Supplier:  VWR Collection
Certificates
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
32 Options Available Below
Product Details & Documents
Test weights in either stainless steel or aluminium.
 : Supplied in a plastic box.
Product Family Options
Aluminium sheet weights with certificate, M2
Product Information
  • Weight
    Tolerance
    Availability
    Price
  • 100 mg
    1,6 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,00 €
    44,00 €
    1 * 1 ST
       44,00 €
  • 200 mg
    2,0 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,00 €
    44,00 €
    1 * 1 ST
       44,00 €
  • 500 mg
    2,5 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,00 €
    44,00 €
    1 * 1 ST
       44,00 €
Stainless steel weights with certificate, M2
Product Information
  • Weight
    Tolerance
    Availability
    Price
  • 1 g
    3 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,90 €
    48,90 €
    1 * 1 ST
       48,90 €
  • 2 g
    4 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,30 €
    49,30 €
    1 * 1 ST
       49,30 €
  • 5 g
    5 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,40 €
    51,40 €
    1 * 1 ST
       51,40 €
  • 10 g
    6 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,80 €
    51,80 €
    1 * 1 ST
       51,80 €
  • 20 g
    8 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,60 €
    52,60 €
    1 * 1 ST
       52,60 €
  • 50 g
    10 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,50 €
    55,50 €
    1 * 1 ST
       55,50 €
  • 100 g
    16  ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,00 €
    56,00 €
    1 * 1 ST
       56,00 €
  • 200 g
    30 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,90 €
    64,90 €
    1 * 1 ST
       64,90 €
  • 500 g
    75 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €
  • 1 kg
    160 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 165,00 €
    165,00 €
    1 * 1 ST
       165,00 €
  • 2 kg
    300 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 232,00 €
    232,00 €
    1 * 1 ST
       232,00 €
  • 5 kg
    750 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 488,00 €
    488,00 €
    1 * 1 ST
       488,00 €
  • 10 kg
    1600 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 861,00 €
    861,00 €
    1 * 1 ST
       861,00 €
Aluminium sheet weights without certificate, M2
Product Information
  • Weight
    Tolerance
    Availability
    Price
  • 100 mg
    1,6 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,50 €
    13,50 €
    1 * 1 ST
       13,50 €
  • 200 mg
    2,0 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,50 €
    13,50 €
    1 * 1 ST
       13,50 €
  • 500 mg
    2,5 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,50 €
    13,50 €
    1 * 1 ST
       13,50 €
Stainless steel weights without certificate, M2
Product Information
  • Weight
    Tolerance
    Availability
    Price
  • 1 g
    3 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,70 €
    16,70 €
    1 * 1 ST
       16,70 €
  • 2 g
    4 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,30 €
    17,30 €
    1 * 1 ST
       17,30 €
  • 5 g
    5 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,10 €
    18,10 €
    1 * 1 ST
       18,10 €
  • 10 g
    6 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,60 €
    20,60 €
    1 * 1 ST
       20,60 €
  • 20 g
    8 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,40 €
    21,40 €
    1 * 1 ST
       21,40 €
  • 50 g
    10 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,20 €
    24,20 €
    1 * 1 ST
       24,20 €
  • 100 g
    16  ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,30 €
    29,30 €
    1 * 1 ST
       29,30 €
  • 200 g
    30 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,10 €
    38,10 €
    1 * 1 ST
       38,10 €
  • 500 g
    75 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 99,70 €
    99,70 €
    1 * 1 ST
       99,70 €
  • 1 kg
    160 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 135,00 €
    135,00 €
    1 * 1 ST
       135,00 €
  • 2 kg
    300 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 207,00 €
    207,00 €
    1 * 1 ST
       207,00 €
  • 5 kg
    750 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 463,00 €
    463,00 €
    1 * 1 ST
       463,00 €
  • 10 kg
    1600 ± mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 839,00 €
    839,00 €
    1 * 1 ST
       839,00 €