Cleanroom sleeve protectors, BioClean-C™
: Nitritex
These chemotherapy sleeve protectors are made of CleanTough™ fabric with 100% polyester cuffs. They are processed to ensure ISO Class 4 compatibility and provide maximum protection against a range of chemotherapy drugs.
- Ultrasonically bonded seams with protective tape
- Lightweight, low linting and hard-wearing
- Elasticated cuffs
- Available sterile or non sterile
Suitable for use in Class 10, ISO 4, M2,5 cleanrooms.
Type PB [6]; Cat. III
: EN ISO 13034:2005 + A1:2010 (Type PB [6]), EN 14325, EN 530, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications.
Sterile model: Sterilised in accordance with ISO 11137-1:2006.
Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications.
Sterile model: Sterilised in accordance with ISO 11137-1:2006.
: Six bags of 15 pairs (90 individually packed pairs) per carton.
- Catalog No:
- 113-8168
- 113-8152
- Cleanroom class:
- 10/ISO 4
- 10/ISO 4
- Garment application:
- Cleanroom, Medical, Laboratory, Clinical
- Cleanroom, Medical, Laboratory, Clinical
Frequently Bought Together