ContactUsLinkComponent
Disposable particle filtering respirators, FFP3, BioClean FFP3™
Disposable particle filtering respirators, FFP3, BioClean FFP3™
Catalog # NITRBFFP3
Supplier:  Nitritex
undefined
Disposable particle filtering respirators, FFP3, BioClean FFP3™
Catalog # NITRBFFP3
Supplier:  Nitritex
Specifications
  • Type:
    Foldable PP mask with headloops
  • Size:
    U*
  • Pk:
    200
  • With valve:
    +
  • Class:
    FFP3
Specifications
About this item
The foldable mask features a one-way valve to reduce exhalation resistance. It offers exceptional respiratory protection by providing 99% filtration of solid particulates and liquid aerosols. The 190×92 mm mask is constructed with white spunbond polypropylene fabric between which is a filter layer of polypropylene meltblown and synthetic fibre composite material. The three layers are ultrasonically sealed.
  • Soft and adjustable aluminium nose band with a soft foam bridge
  • 650 mm synthetic rubber headloop
  • Facial seal made from polyester knitted fabric
Certifications: ISO 9001:2008 and ISO 13485:2003