Order Entry
Austria
ContactUsLinkComponent
Glove box, HEPA-filtered, Precise™
Glove box, HEPA-filtered, Precise™
Catalog # 135-1096
Supplier:  LABCONCO
undefined
Glove box, HEPA-filtered, Precise™
Catalog # 135-1096
Supplier:  LABCONCO

Specifications

  • Volume:
    368,1 L
  • Description:
    Glove box, HEPA-filtered, with 2 glove ports, incl. a pair of neoprene gloves
  • Model:
    Precise™
  • Glove port - int.Ø:
    203 mm
  • Observation window (W×H):
    670×317 mm
  • Transfer chamber W×D×H:
    305 × Ø 279 mm
  • Weight:
    127 kg
  • W×D×H int.:
    851×698×635 mm
  • W×D×H ext.:
    1340×800×1020 mm
  • Pk:
    1

Specifications

About this item

This HEPA-filtered glove box has inlet and outlet HEPA filters and provides a leak-tight physical barrier to protect the operator from hazardous airborne particulates and powders.

  • One–piece, molded polyethylene liner provides a durable surface resistant to chemical corrosion and physical damage while simplifying cleaning with its coved interior corners
  • Dry-powder epoxy-coated steel superstructure, located above the glove box shell for utility placement, has superior corrosion resistance
  • 6,35 mm thick laminated safety glass viewing window, 670×317 mm, removable for loading equipment, with powder-coated steel frame and neoprene gasket
  • Space-saving inner and outer transfer chamber doors that pivot upward, are counterbalanced and equipped with quick-latches
  • Black, chemical-resistant work surface sealed to the glove box floor
  • Built-in blower with speed control capable of airflow from 13 CFM to 100 CFM
  • Inlet and outlet HEPA filters, 99,99% efficient on 0,3 micron particulates

The box is ideal for use in pharmaceutical research, nanotechnology and biochemistry. Microorganisms, low-level radiochemicals, chemical carcinogens and asbestos may also be used in this box. The glove box may be used for cleanroom applications requiring ISO Class 3 (ISO 14644-1) conditions.

Certifications: Meet Class 1 atmosphere containment conditions for oxygen leak rates per ISO 10648-2 test method.
Airborne particulate cleanliness (measured under positive and negative pressures per ISO 14644-1 test method) exceeds ISO Class 5 conditions (Class 100) and achieved ISO Class 3 conditions (Class 1) at both minimum and maximum operational airflow volumes.
Delivery: Supplied with neoprene gloves (size 9 ³/₄").