Inserts for snap ring or crimp neck vials, E-Z Vial®, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Screw caps for certified screw top vials
Supplier: Agilent
Inserts for snap ring or crimp neck vials, E-Z Vial®, WHEATON®
Supplier: DWK Life Sciences
Certified screw top vials and closures
Supplier: Agilent