Test tube half racks, No-Wire™, SP Bel-Art
PP, white
- Steam autoclavable at 121 °C
- Available in 5 grid sizes to accommodate full range of laboratory test tubes
These compact size plastic racks save space in water baths, laminar flow hoods and fume hoods.
Frequently Bought Together