VWR® Traceable®, Waterproof Thermometers
This waterproof and shockproof thermometer is a safe replacement for any hazardous mercury instrument. The probe is resistant to most chemicals.
- Waterproof construction for wet labs, wash-down areas and all field applications
- Memory recalls maximum/minimum temperatures
- Reading updates every second
Stainless steel probe (Lר: 203×3,5 mm).
: Supplied with probe and cover, silver-oxide battery and Traceable® certificate of calibration.
Frequently Bought Together