
1,3,5-Tribenzylhexahydro-1,3,5-triazine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2547-66-2
:
Formula:C₂₄H₂₇N₃
MDL Number:MFCD00014599
:357.5 g/mol
Boiling Pt:100 °C (0.005 mmHg)
Melting Pt:50…51 °C
Flash Pt:113 °C (235 °F)
Storage Temperature:Ambient
EINECS:219-831-7
:GHS07
Related Information
Frequently Bought Together