3-Methylbenzo[b]thiophene-2-carboxaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22053-74-3
:
Formula:C₁₀H₈OS
MDL Number:MFCD00052307
Melting Pt:88…90 °C
:176.24 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together