Order Entry
Austria
ContactUsLinkComponent
Arsenazo III for calcium determination, Sigma-Aldrich®
Toxic
Environment
Arsenazo III for calcium determination, Sigma-Aldrich®
Certificates
undefined
Toxic
Environment
Arsenazo III for calcium determination, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Brown to Very Dark Brown and Black
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    25.5 - 38.1 %
  • Nitrogen:
    5.5 - 8.3 %
  • Calcium (Ca):
    ≤1000 ppm
  • Solubility (Colour)
    c = 1 mg/ml, 1 N NH₄OH:
    Dark Blue to Very Dark Blue and Dark Purple to Very Dark Purple and Blue-Purple

Specification Test Results

Frequently Bought Together