Sodium mercaptoacetate (sodium thioglycolate)
Catalog # MOLE36257729-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: Thioglycolic Acid, Thioglycolic Acid Sodium Salt, Mercaptoacetic acid, sodium salt
:
:114.1 g/mol
EINECS:206-696-4
:GHS07
Storage Temperature:Freezer
CAS Number:367-51-1
Formula:HSCH₂COONa
MDL Number:MFCD00043386