Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Pyruvaldehyde (methylglyoxal) 30-45 (w/w) in water
 
undefined
 
 
 
Pyruvaldehyde (methylglyoxal) 30-45 (w/w) in water

 

  • Appearance:
    Clear yellow to brown Solution
  • Refractive index:
    1.3910 to 1.4200 (20°C, 589 nm)
  • Titration with NaOH:
    30 to 45 %
  • HPLC:
    ≥96.0 %

 

 

Frequently Bought Together