Order Entry
Austria
ContactUsLinkComponent
Catechol violet indicator, Sigma-Aldrich®
Catechol violet indicator, Sigma-Aldrich®
Certificates
undefined
Catechol violet indicator, Sigma-Aldrich®

Specification Test Results

  • Appearance (Form):
    Powder
  • 1H NMR Spectrum:
    Conforms to Structure
  • Solubility (Colour), c = 10 mg/10 ml; Water:
    Faint Brown to Brown and Faint Red to Dark Red
  • Suitability, Suitable for use as an indicator for EDTA titrations:
    Suitable
  • Suitability, Suitable for use as a pH indicator:
    Suitable

Specification Test Results

Frequently Bought Together