Order Entry
Austria
ContactUsLinkComponent
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL, free radical)
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL, free radical)
Catalog # MOLE73749999-5G
Supplier:  Molekula
CAS Number:  
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL, free radical)
Catalog # MOLE73749999-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :172.25 g/mol
EINECS:218-760-9
CAS Number:2226-96-2
Formula:C₉H₁₈NO₂
MDL Number:MFCD00006478