

4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98% free radical
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2226-96-2
:
Formula:C₉H₁₈NO₂
MDL Number:MFCD00006478
Melting Pt:69…72 °C
Flash Pt:146 °C (295 °F)
:172.25 g/mol
EINECS:218-760-9
:GHS05
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together