Order Entry
Austria
ContactUsLinkComponent
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98% free radical
Corrosive
Irritant
Health
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98% free radical
Certificates
undefined
Corrosive
Irritant
Health
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98% free radical

Specification Test Results

  • Appearance:
    Orange Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    69 to 73 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together