Antimony standard solution, 10000 mg/L Sb in water with nitric acid and tartaric acid (each max. 1%) (from Sb), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Sb in H₂O/tartaric acid/tr. HNO₃
Physical Form: Liquid
MDL No.: MFCD00134030
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00134030
Specifications
Related Information
Frequently Bought Together