Order Entry
Austria
ContactUsLinkComponent
Ammonium-Nitrogen standard solution, 100 mg/L NH₄ (as N) in water
Ammonium-Nitrogen standard solution, 100 mg/L NH₄ (as N) in water
Catalog # ACCUICNH4N1X1
Supplier:  AccuStandard
CAS Number:  
Ammonium-Nitrogen standard solution, 100 mg/L NH₄ (as N) in water
Catalog # ACCUICNH4N1X1
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Ammonium-Nitrogen standard solution

Specifications

About this item