Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, p-Phenylenediamine 2,000 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, p-Phenylenediamine 2,000 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERYA16058000AL
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, p-Phenylenediamine 2,000 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERYA16058000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    106-50-3
  • Description:
    p-Phenylenediamine 2,000 µg/ml in acetonitrile
  • Formula:
    C₆H₈N₂
  • Description:
    p-Phenylenediamine

Specifications

About this item