Order Entry
Austria
ContactUsLinkComponent
Boron standard solution, 100 mg/L B in water (from B(OH₃))
Boron standard solution, 100 mg/L B in water (from B(OH₃))
Catalog # CMPLFC31.K1.W.L25
CAS Number:  
Boron standard solution, 100 mg/L B in water (from B(OH₃))
Catalog # CMPLFC31.K1.W.L25
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item